R&D Systems
Manufacturer Code:FAB1621N100UG
Catalog # FAB1621N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 120612 |
Immunogen | NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGNR Accession # Q9H2X3 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; CD209; CD209 antigen; DC-SIGN; Dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein
Gene Aliases: CD209; CDSIGN; CLEC4L; DC-SIGN; DC-SIGN1
UniProt ID: (Human) Q9NNX6
Entrez Gene ID: (Human) 30835
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.