Invitrogen
Manufacturer Code:MA140070
Catalog # 01-676-508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | ER-TR9 |
Immunogen | Murine SIGN-R1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD209; CD209 antigen-like protein A; CD209 CDSIGN CIRE DC-SIGN DC-SIGN1 Dcsign MGC130443 SIGN-R1 SIGNR5 CD209a; DC-SIGN; Dendritic cell-specific ICAM-3-grabbing non-integrin
Gene Aliases: CD209; Cd209a; CDSIGN; CIRE; DC-SIGN; DC-SIGN1; Dcsign; SIGN-R1; SIGNR5
UniProt ID: (Mouse) Q91ZX1
Entrez Gene ID: (Mouse) 170786
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.