Invitrogen
Manufacturer Code:MA535983
Catalog # PIMA535983
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 037 |
Immunogen | Recombinant Human CD209/DC-SIGN Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; CD209; CD209 antigen; CD209 CD209 antigen CD209 antigen-like protein A CD209 molecule Cd209a CD209a antigen CD209a molecule Cd209d CD209d antigen CDSIGN Cire CLEC4L Clec4m C-type lectin domain family 4 member L C-type lectin domain family 4 member L C-type lectin domain family 4 member M Dcsign DC-SIGN DC-SIGN1 Dc-signr DC-SIGN-related protein Dendritic cell-specific ICAM-3-grabbing non-integrin Dendritic cell-specific ICAM-3-grabbing non-integrin 1 dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin HIV gpl20-binding protein MGC129965 MGC130443 RGD1561104 SIGN-R1 SIGNR5; DC-SIGN; Dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein
Gene Aliases: CD209; CDSIGN; CLEC4L; DC-SIGN; DC-SIGN1
UniProt ID: (Human) Q9NNX6
Entrez Gene ID: (Human) 30835
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.