Invitrogen
Manufacturer Code:PA587027
Catalog # PIPA587027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human CD209. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; CD209; CD209 antigen; CD209 antigen-like protein A; CD209a antigen; CD209a molecule; CDSIGN CLEC4L DC-SIGN DC-SIGN1; DC-SIGN; Dendritic cell-specific ICAM-3-grabbing non-integrin; Dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein
Gene Aliases: CD209; Cd209a; CDSIGN; CIRE; CLEC4L; DC-SIGN; DC-SIGN1; Dcsign; SIGN-R1; SIGNR5
UniProt ID: (Human) Q9NNX6, (Mouse) Q91ZX1
Entrez Gene ID: (Human) 30835, (Rat) 288375, (Mouse) 170786
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.