Invitrogen
Manufacturer Code:PA538237
Catalog # PIPA538237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from CDEDEEGEEDEEITVS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Developing brain homeobox protein 1; Homeobox protein DBX1; Mmox C
Gene Aliases: AI426026; Dbx; DBX1
UniProt ID: (Human) A6NMT0, (Mouse) P52950, (Rat) Q5NSW5
Entrez Gene ID: (Human) 120237, (Mouse) 13172, (Rat) 292934
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.