Novus Biologicals
Manufacturer Code:NBP297356FR
Catalog # NB042679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human DBR1 fragment. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: debranching enzyme (S. Cerevisiae) homolog 1 debranching enzyme homolog 1 (S. cerevisiae) EC 3.1 lariat debranching enzyme RNA lariat debranching enzyme; debranching enzyme homolog 1; Lariat debranching enzyme; RNA lariat debranching enzyme
Gene Aliases: DBR1
UniProt ID: (Human) Q9UK59
Entrez Gene ID: (Human) 51163
Molecular Function: RNA binding protein endoribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.