Proteintech
Manufacturer Code:160191AP150UL
Catalog # 50-559-296
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DBR1 Fusion Protein Ag8830 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DBR1 Lariat debranching enzyme; debranching enzyme homolog 1; Lariat debranching enzyme; RNA lariat debranching enzyme
Gene Aliases: AW018415; DBR1
UniProt ID: (Human) Q9UK59, (Mouse) Q923B1
Entrez Gene ID: (Human) 51163, (Rat) 681234, (Mouse) 83703
Molecular Function:
RNA binding protein
endoribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.