Abnova
Manufacturer Code:H00001628M01
Catalog # 89-021-262
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A6 |
Immunogen | DBP (NP_001343 226 a.a. ~ 325 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant DBP.
For Research Use Only
Protein Aliases: Albumin D box-binding protein; Albumin D-element-binding protein; D site of albumin promoter (albumin D-box) binding protein; D site-binding protein; DABP; Tax-responsive enhancer element-binding protein 302; TaxREB302
Gene Aliases: DABP; DBP
UniProt ID: (Human) Q10586
Entrez Gene ID: (Human) 1628
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.