Invitrogen
Manufacturer Code:PA540501
Catalog # PIPA540501
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human DBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Albumin D box-binding protein; albumin D-element binding protein; Albumin D-element-binding protein; D site albumin promoter binding protein; D site albumin promoter-binding protein 1; D site of albumin promoter (albumin D-box) binding protein; D site of albumin promoter binding protein; D site-binding protein; DBP; Tax-responsive enhancer element-binding protein 302; TaxREB302
Gene Aliases: DABP; DBP
UniProt ID: (Bovine) Q32PF6, (Human) Q10586, (Rat) P16443, (Mouse) Q60925
Entrez Gene ID: (Bovine) 503577, (Horse) 100146301, (Human) 1628, (Dog) 100685867, (Sheep) 100192428, (Rat) 24309, (Mouse) 13170
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.