Proteintech
Manufacturer Code:130151AP150UL
Catalog # 50-172-8421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DBNL Fusion Protein Ag4078 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Abp1; ABP1 Cervical SH3P7 CMAP DBNL Drebrin F drebrin like Drebrin like protein HIP 55 SH3P7; Actin-binding protein 1; Cervical mucin-associated protein; Cervical SH3P7; dbnl; dbnl {ECO:0000312|EMBL:AAH72483.1}; Drebrin-F; drebrin-like; Drebrin-like protein; HIP-55; HPK1-interacting protein of 55 kDa; SH3 domain-containing protein 7; src homology 3 domain-containing protein HIP-55
Gene Aliases: ABP1; CMAP; DBNL; HIP-55; HIP55; mAbp1; PP5423; SH3P7
UniProt ID: (Human) Q9UJU6, (Rat) Q9JHL4, (Mouse) Q62418
Entrez Gene ID: (Human) 28988, (Rat) 83527, (Mouse) 13169
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.