Novus Biologicals
Manufacturer Code:NBP2827970.1ML
Catalog # NB030726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human DBNDD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf35 chromosome 20 open reading frame 35 CK1BP dysbindin (dystrobrevin binding protein 1) domain containing 2 dysbindin domain-containing protein 2 HSMNP1Casein kinase-1 binding protein SCF apoptosis response protein 1; Casein kinase-1 binding protein; CK1BP; dysbindin (dystrobrevin binding protein 1) domain containing 2; Dysbindin domain-containing protein 2; HSMNP1; SCF apoptosis response protein 1
Gene Aliases: C20orf35; CK1BP; DBNDD2; HSMNP1
UniProt ID: (Human) Q9BQY9
Entrez Gene ID: (Human) 55861
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.