Proteintech
Manufacturer Code:276231AP150UL
Catalog # 50-173-5718
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DBNDD2 Fusion Protein Ag26356 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C20orf35 CK1BP DBNDD2 HSMNP1; Casein kinase-1 binding protein; CK1BP; dysbindin (dystrobrevin binding protein 1) domain containing 2; Dysbindin domain-containing protein 2; HSMNP1; likely orthologue of H. sapiens chromosome 20 open reading frame 35; SCF apoptosis response protein 1
Gene Aliases: 1110017A21Rik; 2900022J10Rik; AU041050; AW048677; C20orf35; CK1BP; D2Bwg0891e; DBNDD2; HSMNP1; Nkip; R74724
UniProt ID: (Human) Q9BQY9, (Mouse) Q9CRD4
Entrez Gene ID: (Human) 55861, (Mouse) 52840, (Rat) 499941
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.