Proteintech
Manufacturer Code:144901AP150UL
Catalog # 50-172-9486
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DBI Fusion Protein Ag5891 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACBD1 ACBP Acyl CoA binding protein CCK RP DBI Diazepam binding inhibitor Endozepine EP; ACBP; acyl coenzyme A binding protein; acyl-CoA binding protein; Acyl-CoA-binding protein; acyl-Coenzyme A binding domain containing 1; cholecystokinin-releasing peptide, trypsin-sensitive; DBI; Diazepam binding inhibitor (GABA receptor modulator acyl-Coenxyme A binding protein); diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein); Diazepam binding inhibitor (GABA receptor modulator, acyl-Coenxyme A binding protein); diazepam binding inhibitor (GABA receptor modulator, acyl-Coenzyme A binding protein); diazepam binding inhibitor, splice form 1b; Diazepam-binding inhibitor; Endozepine; EP; GABA receptor modulator; LRRGT00046; Octadecaneuropeptide; ODN; Triakontatetraneuropeptide; TTN
Gene Aliases: ACBD1; ACBP; Acoabp3; CCK-RP; DBI; endozepine; EP; Odn; RNACOABP3; Ttn
UniProt ID: (Human) P07108, (Mouse) P31786, (Rat) P11030
Entrez Gene ID: (Human) 1622, (Mouse) 13167, (Rat) 25045
Molecular Function:
enzyme modulator
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.