Abnova
Manufacturer Code:PAB9692
Catalog # 89-134-338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus human DBH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Sheep polyclonal antibody raised against synthetic peptide of DBH.
For Research Use Only
Protein Aliases: DBM; Dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); Dopamine beta-monooxygenase; Soluble dopamine beta-hydroxylase
Gene Aliases: DBH; DBM
UniProt ID: (Human) P09172
Entrez Gene ID: (Human) 1621
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.