Abnova
Manufacturer Code:PAB14325
Catalog # 89-117-004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Inhibition Assays (IA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Radioimmune Assays (RIA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified DBH from bovine adrenal glands. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with deionized water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against native DBH.
For Research Use Only
Protein Aliases: Dopamine beta-hydroxylase; dopamine beta-hydroxylase precursor (EC 1.14.17.1); Dopamine beta-monooxygenase; dopamine beta-monooxygenase precursor (EC 1.14.17.1); mixed function oxidase; Soluble dopamine beta-hydroxylase
Gene Aliases: DBH
UniProt ID: (Bovine) P15101
Entrez Gene ID: (Bovine) 280758
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.