Invitrogen
Manufacturer Code:PA571078
Catalog # PIPA571078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human DBF4B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activator of S phase kinase-like protein 1; activator of S-phase kinase-like protein 1; ASK-like protein 1; ASKL1 CHIFB DRF1 ZDBF1B ASK-like protein 1 Dbf4-related factor 1 activator of S-phase kinase-like protein 1 chiffon homolog B protein DBF4 homolog B zinc finger DBF-type containing 1B; Chiffon homolog B; DBF4 homolog B; Dbf4-related factor 1; Protein DBF4 homolog B; zinc finger, DBF-type containing 1B
Gene Aliases: ASKL1; CHIFB; DBF4B; DRF1; ZDBF1B
UniProt ID: (Human) Q8NFT6
Entrez Gene ID: (Human) 80174
Molecular Function: enzyme modulator kinase activator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.