Novus Biologicals
Manufacturer Code:NBP213900
Catalog # NBP213900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: HPSQENSFAPADIPVKGPLLFPEARPWLMSARCWVRPFPFVTWGCLIPHD TTPLHEEVSPCPCLRLGYLYLLLTQSLWCRVRVPSLSTAGPIPRTSHPCT LAFPSYLNDHDLGHLCQAKPQGWNTPQPFLHCGFLA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activator of S phase kinase-like protein 1; Activator of S phase kinase-like protein 1 activator of S-phase kinase-like protein 1 ASKL1CHIFB ASK-like protein 1 chifb Chiffon homolog B DBF4 homolog B (S. cerevisiae) Dbf4-related factor 1 DRF1MGC15009 FLJ13087 protein DBF4 homolog B ZDBF1B zinc finger DBF-type containing 1B; activator of S-phase kinase-like protein 1; ASK-like protein 1; Chiffon homolog B; DBF4 homolog B; Dbf4-related factor 1; Protein DBF4 homolog B; zinc finger, DBF-type containing 1B
Gene Aliases: ASKL1; CHIFB; DBF4B; DRF1; ZDBF1B
UniProt ID: (Human) Q8NFT6
Entrez Gene ID: (Human) 80174
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.