Invitrogen
Manufacturer Code:PA537959
Catalog # PIPA537959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-ESHNRFSATEVTNK from the internal region of CCAR2 (aa842-855) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cell cycle and apoptosis regulator 2 DBC-1 KIAA1967 NET35 p30 DBC p30DBC cell cycle and apoptosis regulator protein 2 cell division cycle and apoptosis regulator protein 2 deleted in breast cancer 1 p30 DBC protein; Cell cycle and apoptosis regulator protein 2; Cell division cycle and apoptosis regulator protein 2; DBC-1; DBIRD complex subunit KIAA1967; deleted in breast cancer 1; Deleted in breast cancer gene 1 protein; NET35; p30 DBC; p30 DBC protein
Gene Aliases: 2610301G19Rik; CCAR2; DBC-1; DBC1; KIAA1967; mKIAA1967; NET35; p30 DBC; p30DBC
UniProt ID: (Human) Q8N163, (Mouse) Q8VDP4
Entrez Gene ID: (Human) 57805, (Mouse) 219158
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.