Invitrogen
Manufacturer Code:PA5101202
Catalog # PIPA5101202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KIAA1967 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610301G19Rik Ccar2 cell cycle activator and apoptosis regulator 2 cell cycle and apoptosis regulator 2 cell cycle and apoptosis regulator protein 2 Cell division cycle and apoptosis regulator protein 2 DBC.1 DBC1 DBC-1 DBCCR1 DBIRD complex subunit KIAA1967 deleted in breast cancer 1 Deleted in breast cancer gene 1 protein FAM5A KIAA1967 mKIAA1967 NET35 p30 DBC p30 DBC protein p30DBC RGD1309922; Cell cycle and apoptosis regulator protein 2; Cell division cycle and apoptosis regulator protein 2; DBC-1; DBIRD complex subunit KIAA1967; deleted in breast cancer 1; Deleted in breast cancer gene 1 protein; NET35; p30 DBC; p30 DBC protein
Gene Aliases: CCAR2; DBC-1; DBC1; KIAA1967; NET35; p30 DBC; p30DBC
UniProt ID: (Human) Q8N163
Entrez Gene ID: (Human) 57805
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.