Invitrogen
Manufacturer Code:PA523152
Catalog # PIPA523152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (50 RMDETEIRSFFARYG 64) of DAZL protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAZ homolog; DAZ-like autosomal; DAZH DAZL1 DAZLA SPGYLA DAZ homolog DAZ-like autosomal SPGY-like-autosomal deleted in azoospermia-like 1 germline specific RNA binding protein spermatogenesis gene on the Y-like autosomal; Deleted in azoospermia-like; Deleted in azoospermia-like 1; deleted in azoospermia-like protein; germline specific RNA binding protein; spermatogenesis gene on the Y-like autosomal; SPGY-like-autosomal; testis secretory sperm-binding protein Li 204a
Gene Aliases: Daz-like; DAZH; DAZL; DAZL1; DAZLA; SPGYLA; Tpx-2; Tpx2
UniProt ID: (Human) Q92904, (Mouse) Q64368
Entrez Gene ID: (Human) 1618, (Dog) 477055, (Bovine) 530116, (Mouse) 13164
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.