Novus Biologicals
Manufacturer Code:NBP185307
Catalog # NBP185307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PTYPNSPVQVITGYQLPVYNYQMPPQWPVGEQRSYVVPPAYSAVNYHCNEVDPGAEVVPNECS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAZ homolog; DAZ-like autosomal; DAZHSPGY-like-autosomal DAZL1DAZ-like autosomal DAZLA deleted in azoospermia-like Deleted in azoospermia-like 1 germline specific RNA binding protein MGC26406 spermatogenesis gene on the Y-like autosomal SPGYLADAZ homolog; Deleted in azoospermia-like; Deleted in azoospermia-like 1; germline specific RNA binding protein; spermatogenesis gene on the Y-like autosomal; SPGY-like-autosomal; testis secretory sperm-binding protein Li 204a
Gene Aliases: DAZH; DAZL; DAZL1; DAZLA; SPGYLA
UniProt ID: (Human) Q92904
Entrez Gene ID: (Human) 1618
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.