Abnova
Manufacturer Code:PAB6834
Catalog # 89-132-724
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human DAZL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of DAZL.
For Research Use Only
Protein Aliases: DAZ homolog; DAZ-like autosomal; DAZH; DAZL1; DAZLA; Deleted in azoospermia-like; Deleted in azoospermia-like 1; germline specific RNA binding protein; MGC26406; spermatogenesis gene on the Y-like autosomal; SPGY-like-autosomal; SPGYLA; testis secretory sperm-binding protein Li 204a
Gene Aliases: DAZH; DAZL; DAZL1; DAZLA; SPGYLA
UniProt ID: (Human) Q92904
Entrez Gene ID: (Human) 1618
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.