Invitrogen
Manufacturer Code:PA565420
Catalog # PIPA565420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human DAZAP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAZ-associated protein 2; deleted in azoospermia associated protein 2; Deleted in azoospermia-associated protein 2; proline-rich transcript in brain; proline-rich transcript, brain-expressed protein; PRTB DAZ-associated protein 2 deleted in azoospermia associated protein 2 deleted in azoospermia-associated protein 2 proline-rich transcript in brain proline-rich transcript brain-expressed protein
Gene Aliases: DAZAP2; KIAA0058; PRTB
UniProt ID: (Human) Q15038
Entrez Gene ID: (Human) 9802
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.