Invitrogen
Manufacturer Code:PA552074
Catalog # PIPA552074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DAZAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAZ-associated protein 1; DAZ-associated protein 1 Deleted in azoospermia-associated protein 1 DAZAP1; Deleted in azoospermia-associated protein 1; testicular tissue protein Li 50
Gene Aliases: 2410042M16Rik; DAZAP1; mPrrp
UniProt ID: (Human) Q96EP5, (Mouse) Q9JII5
Entrez Gene ID: (Human) 26528, (Mouse) 70248, (Rat) 362836
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.