Invitrogen
Manufacturer Code:PA541886
Catalog # PIPA541886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human DAZAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAZ associated protein 1; DAZ-associated protein 1; Deleted in azoospermia-associated protein 1; testicular tissue protein Li 50
Gene Aliases: 2410042M16Rik; DAZAP1; mPrrp
UniProt ID: (Human) Q96EP5, (Mouse) Q9JII5
Entrez Gene ID: (Dog) 100683271, (Horse) 100068825, (Human) 26528, (Mouse) 70248, (Rat) 362836, (Guinea pig) 100732237, (Zebrafish) 561459
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.