Novus Biologicals
Manufacturer Code:NBP15725520UL
Catalog # NBP15725520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to DAZAP1 (DAZ associated protein 1) The peptide sequence was selected from the C terminal of DAZAP1. Peptide sequence GFGQGFSDPSQQPPSYGGPSVPGSGGPPAGGSGFGRGQNHNVQGFHPYRR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAZ associated protein 1 DAZ-associated protein 1 deleted in azoospermia associated protein 1 Deleted in azoospermia-associated protein 1 MGC19907; DAZ-associated protein 1; Deleted in azoospermia-associated protein 1; testicular tissue protein Li 50
Gene Aliases: DAZAP1
UniProt ID: (Human) Q96EP5
Entrez Gene ID: (Human) 26528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.