Invitrogen
Manufacturer Code:PA579137
Catalog # PIPA579137
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human Daxx recombit protein (Position: K56-R345). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CENP-C binding protein; Daxx; Death domain-associated protein 6; Death domain-associated protein 6 Daxx hDaxx ETS1-associated protein 1 EAP1 Fas death domain-associated protein DAXX BING2 DAP6; death-associated protein 6; death-domain associated protein; EAP1; ETS1-associated protein 1; Fas death domain-associated protein; Fas-binding protein; hDaxx
Gene Aliases: BING2; DAP6; DAXX; EAP1
UniProt ID: (Human) Q9UER7
Entrez Gene ID: (Human) 1616, (Rat) 140926
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.