Novus Biologicals
Manufacturer Code:NBP270116AF488
Catalog # NB270116A48
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F5 |
Immunogen | Recombinant protein fragment corresponding to amino acids 161-430 of human DAX1/NR0B1 (NP_000466) produced in E.coli. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AHCDSS AHCH AHX DAX1DAX-1 dosage-sensitive sex reversal DSS-AHC critical region on the X chromosome protein 1 GTD HHG NROB1 nuclear hormone receptor Nuclear receptor DAX-1 nuclear receptor subfamily 0 group B member 1 nuclear receptor subfamily 0 group B member 1; DSS-AHC critical region on the X chromosome protein 1; nuclear hormone receptor; Nuclear receptor DAX-1; Nuclear receptor subfamily 0 group B member 1; nuclear receptor subfamily 0, group B, member 1
Gene Aliases: AHC; AHCH; AHX; DAX-1; DAX1; DSS; GTD; HHG; NR0B1; NROB1; SRXY2
UniProt ID: (Human) P51843
Entrez Gene ID: (Human) 190
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.