R&D Systems
Manufacturer Code:PPH743100
Catalog # PPH743100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | H7431 |
Immunogen | Recombinant human DAX1 (amino acids 1 - 80) and mouse myeloma cells (NS-1) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AHC AHCDSS AHCH AHX DAX1DAX-1 dosage-sensitive sex reversal DSS-AHC critical region on the X chromosome protein 1 GTD HHG NR0B1 NROB1 nuclear hormone receptor Nuclear receptor DAX-1 nuclear receptor subfamily 0 group B member 1 nuclear receptor subfamily 0 group B member 1; DSS-AHC critical region on the X chromosome protein 1; nuclear hormone receptor; Nuclear receptor DAX-1; Nuclear receptor subfamily 0 group B member 1; nuclear receptor subfamily 0, group B, member 1
Gene Aliases: AHC; AHCH; AHX; DAX-1; DAX1; DSS; GTD; HHG; NR0B1; NROB1; SRXY2
UniProt ID: (Human) P51843
Entrez Gene ID: (Human) 190
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.