Invitrogen
Manufacturer Code:PA5114321
Catalog # PIPA5114321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein within Zebrafish DARS2 aa 43-321. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5830468K18Rik aspartate tRNA ligase 2 mitochondrial aspartate--tRNA ligase mitochondrial Aspartyl-tRNA synthetase aspartyl-tRNA synthetase 2 (mitochondrial) aspartyl-tRNA synthetase 2 mitochondrial aspartyl-tRNA synthetase mitochondrial aspRS Dars2 LBSL MT-ASPRS RGD1308286
Gene Aliases: si:dkey-21n10.1
Entrez Gene ID: (Zebrafish) 571144
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.