Novus Biologicals
Manufacturer Code:NBP183854
Catalog # NBP183854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LSKPHGTVKAICIPEGAKYLKRKDIESIRNFAADHFNQEILPVFLNANRNWNSPVANFIMESQRLELIRLMETQEEDVVLLTA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aspartate tRNA ligase 2 mitochondrial Aspartate--tRNA ligase aspartyl-tRNA synthetase 2 mitochondrial aspartyl-tRNA synthetase mitochondrial AspRS ASPRS. LBSL EC 6.1.1 EC 6.1.1.12 FLJ10514 LBSL MT-ASPRS RP3-383J4.2; aspartate tRNA ligase 2, mitochondrial; Aspartate--tRNA ligase, mitochondrial; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, mitochondrial; AspRS
Gene Aliases: ASPRS; DARS2; LBSL; MT-ASPRS
UniProt ID: (Human) Q6PI48
Entrez Gene ID: (Human) 55157
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.