Proteintech
Manufacturer Code:138071AP150UL
Catalog # 50-172-9018
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DARS2 Fusion Protein Ag4809 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aspartate tRNA ligase 2, mitochondrial; Aspartate tRNA ligase ASPRS DARS2 LBSL MT ASPRS RP3 383J4.2; Aspartate--tRNA ligase, mitochondrial; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, mitochondrial; AspRS
Gene Aliases: 5830468K18Rik; ASPRS; DARS2; LBSL; MT-ASPRS; RGD1308286
UniProt ID: (Human) Q6PI48, (Rat) Q3KRD0, (Mouse) Q8BIP0
Entrez Gene ID: (Human) 55157, (Rat) 304919, (Mouse) 226539
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.