Invitrogen
Manufacturer Code:PA556266
Catalog # PIPA556266
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aspartate tRNA ligase 1, cytoplasmic; Aspartate--tRNA ligase, cytoplasmic; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, cytoplasmic; AspRS; aspRS aspartate tRNA ligase 1 cytoplasmic aspartate--tRNA ligase cytoplasmic aspartyl-tRNA synthetase cytoplasmic cell proliferation-inducing gene 40 protein cell proliferation-inducing protein 40; Cell proliferation-inducing gene 40 protein; testicular tissue protein Li 192
Gene Aliases: 5730439G15Rik; aspRS; DARS; DARS1; HBSL; PIG40
UniProt ID: (Human) P14868, (Mouse) Q922B2
Entrez Gene ID: (Human) 1615, (Mouse) 226414
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.