Novus Biologicals
Manufacturer Code:NBP300268G
Catalog # NB087102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant DARS (P14868 Met1-Pro501). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aspartate tRNA ligase 1 cytoplasmic Aspartate--tRNA ligase aspartyl-tRNA synthetase aspartyl-tRNA synthetase cytoplasmic aspRS Cell proliferation-inducing gene 40 protein cell proliferation-inducing protein 40 DKFZp781B11202 EC 6.1.1.12 MGC111579; aspartate tRNA ligase 1, cytoplasmic; Aspartate--tRNA ligase, cytoplasmic; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, cytoplasmic; AspRS; Cell proliferation-inducing gene 40 protein; testicular tissue protein Li 192
Gene Aliases: aspRS; DARS; DARS1; HBSL; PIG40
UniProt ID: (Human) P14868
Entrez Gene ID: (Human) 1615
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.