R&D Systems
Manufacturer Code:MAB4230
Catalog # MAB4230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 375604 |
Immunogen | <EM>E. coli</EM>-derived recombinant human DARPP-32 Ala51-Thr204 Accession # Q9UD71.2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: DARPP-32; DARPP32 PPP1R1B protein phosphatase 1 regulatory (inhibitor) subunit 1B regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein DARPP-32); dopamine and cAMP-regulated neuronal phosphoprotein 32; Dopamine- and cAMP-regulated neuronal phosphoprotein; Protein phosphatase 1 regulatory subunit 1B
Gene Aliases: DARPP-32; DARPP32; PPP1R1B
UniProt ID: (Human) Q9UD71
Entrez Gene ID: (Human) 84152
Molecular Function:
enzyme modulator
phosphatase inhibitor
phosphatase modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.