Novus Biologicals
Manufacturer Code:NBP238908
Catalog # NBP238908
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: EEEEDSQAEVLKVIRQSAGQKTTCGQGLEGPWERPPPLDESERDGGSEDQVEDPALSEPG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DARPP-32; dopamine and cAMP-regulated neuronal phosphoprotein 32; Dopamine- and cAMP-regulated neuronal phosphoprotein; protein phosphatase 1 regulatory (inhibitor) subunit 1B regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein DARPP-32); Protein phosphatase 1 regulatory subunit 1B
Gene Aliases: DARPP-32; DARPP32; PPP1R1B
UniProt ID: (Human) Q9UD71
Entrez Gene ID: (Human) 84152
Molecular Function:
enzyme modulator
phosphatase inhibitor
phosphatase modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.