R&D Systems
Manufacturer Code:FAB4139V100UG
Catalog # FAB4139V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 358307 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human DARC Met1-Ser336 Accession # Q16570 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: ACKR1 CCBP1 CD234 CD234 antigen Dfy Duffy Antigen Duffy antigen/chemokine receptor Duffy blood group antigen Duffy blood group chemokine receptor Fy Antigen Fy glycoprotein FYDuffy blood group Glycoprotein D GPD GPDWBCQ1 GpFy Plasmodium vivax receptor; Atypical chemokine receptor 1; CD234; Duffy antigen chemokine receptor; Duffy antigen/chemokine receptor; Duffy blood group antigen; duffy blood group chemokine receptor; Duffy blood group, atypical chemokine receptor; Duffy blood group, chemokine receptor; Fy glycoprotein; Glycoprotein D; GpFy; Plasmodium vivax receptor
Gene Aliases: ACKR1; CCBP1; CD234; DARC; Dfy; FY; GPD; GpFy; WBCQ1
UniProt ID: (Human) Q9UQE1
Entrez Gene ID: (Human) 2532
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.