Novus Biologicals
Manufacturer Code:NBP275197
Catalog # NBP275197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Blocking Assay (BLOCK) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2C3 |
Immunogen | CHO cells transfected with cDNA encoding the Duffy glycoprotein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for up to 3 months. For longer storage aliquot and store at -20°C. |
Rabbit Monoclonal Antibody
Protein Aliases: Atypical chemokine receptor 1; CCBP1 CD234 CD234 antigen Dfy Duffy antigen/chemokine receptor Duffy blood group antigen Duffy blood group chemokine receptor Fy glycoprotein FYDuffy blood group Glycoprotein D GPDWBCQ1 GpFy Plasmodium vivax receptor; CD234; Duffy antigen chemokine receptor; Duffy antigen/chemokine receptor; Duffy blood group antigen; duffy blood group chemokine receptor; Duffy blood group, atypical chemokine receptor; Duffy blood group, chemokine receptor; Fy glycoprotein; Glycoprotein D; GpFy; Plasmodium vivax receptor
Gene Aliases: ACKR1; CCBP1; CD234; DARC; Dfy; FY; GPD; GpFy; WBCQ1
UniProt ID: (Human) Q9UQE1
Entrez Gene ID: (Human) 2532
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.