R&D Systems
Manufacturer Code:FAB6695P
Catalog # FAB6695P
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombinant mouse DARC Met1-Pro61 Ala115-Cys127 Ser186-Lys205 Tyr264-Asn285 Accession # NP_034175 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: ACKR1 CCBP1 CD234 CD234 antigen Dfy Duffy Antigen Duffy antigen/chemokine receptor Duffy blood group antigen Duffy blood group chemokine receptor Fy Antigen Fy glycoprotein FYDuffy blood group Glycoprotein D GPD GPDWBCQ1 GpFy Plasmodium vivax receptor; Atypical chemokine receptor 1; CD234; Duffy antigen chemokine receptor; Duffy antigen/chemokine receptor; Duffy blood group antigen; duffy blood group chemokine receptor; Duffy blood group, atypical chemokine receptor; Duffy blood group, chemokine receptor; Fy glycoprotein; Glycoprotein D; GpFy; Plasmodium vivax receptor
Gene Aliases: ACKR1; CCBP1; CD234; DARC; Dfy; FY; GPD; GpFy; WBCQ1
UniProt ID: (Human) Q9UQE1
Entrez Gene ID: (Human) 2532
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.