Proteintech
Manufacturer Code:551851AP150UL
Catalog # 50-173-6043
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Atypical chemokine receptor 1; CCBP1 CD234 DARC Dfy FY Fy glycoprotein Glycoprotein D GPD GpFy Plasmodium vivax receptor WBCQ1; CD234; Duffy antigen chemokine receptor; Duffy antigen receptor for chemokines; Duffy antigen/chemokine receptor; Duffy blood group antigen; duffy blood group chemokine receptor; Duffy blood group, atypical chemokine receptor; Duffy blood group, chemokine receptor; Fy glycoprotein; Glycoprotein D; GpFy; Plasmodium vivax receptor
Gene Aliases: AA162249; ACKR1; CCBP1; CD234; DARC; Dfy; ESTM35; FY; GPD; GpFy; WBCQ1
UniProt ID: (Human) Q9UQE1, (Mouse) Q9QUI6
Entrez Gene ID: (Human) 2532, (Rat) 689105, (Mouse) 13349
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.