R&D Systems
Manufacturer Code:AF7024
Catalog # AF7024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human DAPP1 Gly2-Ser163 Accession # Q9UN19 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: B lymphocyte adapter protein Bam32; B lymphocyte adapter protein Bam32 BAM32 BAM32DKFZp667E0716 B-cell adapter molecule of 32 kDa dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide dual adaptor of phosphotyrosine and 3-phosphoinositides hDAPP1; B-cell adapter molecule of 32 kDa; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; hDAPP1
Gene Aliases: BAM32; DAPP1; HSPC066
UniProt ID: (Human) Q9UN19
Entrez Gene ID: (Human) 27071
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.