Proteintech
Manufacturer Code:147221AP150UL
Catalog # 50-172-9672
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DAPP1 Fusion Protein Ag6459 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B lymphocyte adapter protein Bam32; B-cell adapter molecule of 32 kDa; BAM32 DAPP1 hDAPP1; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; dual adaptor for phosphotyrosine and 3-phosphoinositides 1; hDAPP1; mDAPP1
Gene Aliases: BAM32; DAPP1; HSPC066
UniProt ID: (Human) Q9UN19, (Mouse) Q9QXT1
Entrez Gene ID: (Human) 27071, (Mouse) 26377, (Rat) 362046
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.