OriGene
Manufacturer Code:CF501086
Catalog # 50-166-0124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F7 |
Immunogen | Full length human recombit protein of human DAPK2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DAP kinase 2; DAP-kinase-related protein 1; DAP-kinase-related protein 1 beta isoform; Death-associated protein kinase 2; DRP-1; DRP-1 DRP1
Gene Aliases: DAPK2; DRP-1; DRP1
UniProt ID: (Human) Q9UIK4
Entrez Gene ID: (Human) 23604
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.