Invitrogen
Manufacturer Code:MA534929
Catalog # PIMA534929
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 12B2 |
Immunogen | Recombinant protein within Human DAP Kinase 1 aa 450-670. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: D13Ucla1 DAP kinase 1 DAPK DAPK1 DAP-Kinase Death-associated protein kinase 1 DKFZp781I035 OTTHUMP00000216193; DAP kinase 1; Death-associated protein kinase 1
Gene Aliases: DAPK; DAPK1
UniProt ID: (Human) P53355
Entrez Gene ID: (Human) 1612
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.