Novus Biologicals
Manufacturer Code:NB12011929UV
Catalog # NB12011929U
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 39C534.1 |
Immunogen | The antibody was raised against two synthetic peptides corresponding to amino acids 796-812 of human DAP-5 protein. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: aging-associated protein 1; aging-associated protein 1 DAP-5 DAP5AAG1 Death-associated protein 5 eIF4G 2 eIF-4G 2 eIF-4-gamma 2 eukaryotic translation initiation factor 4 gamma 2 eukaryotic translation initiation factor 4 gamma 2 eukaryotic translation initiation factor 4G-like 1 FLJ41344 NAT1 p97; DAP-5; Death-associated protein 5; eIF-4-gamma 2; eIF-4G 2; eIF4G 2; EIF4G2; Eukaryotic translation initiation factor 4 gamma 2; eukaryotic translation initiation factor 4 gamma, 2; eukaryotic translation initiation factor 4G-like 1; p97
Gene Aliases: AAG1; DAP5; EIF4G2; NAT1; OK/SW-cl.75; P97
UniProt ID: (Human) P78344
Entrez Gene ID: (Human) 1982
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.