Invitrogen
Manufacturer Code:PA521377
Catalog # PIPA521377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 194 (P78344) of DAP5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aging-associated protein 1; DAP-5; Death-associated protein 5; eIF-4-gamma 2; eIF-4G 2; eIF4G 2; EIF4G2; Eukaryotic translation initiation factor 4 gamma 2; eukaryotic translation initiation factor 4 gamma, 2; eukaryotic translation initiation factor 4G-like 1; Novel APOBEC-1 target 1; OK/SW-cl.75 AAG1 DAP5 NAT1 P97 DAP-5 aging-associated protein 1 death-associated protein 5 eIF-4-gamma 2 eIF-4G 2 eIF4G 2 eukaryotic translation initiation factor 4 gamma 2 eukaryotic translation initiation factor 4G-like 1; p97; Translation repressor NAT1
Gene Aliases: AA589388; AAG1; DAP-5; DAP5; E130105L11Rik; EIF4G2; NAT1; Natm1; OK/SW-cl.75; P97
UniProt ID: (Human) P78344, (Mouse) Q62448
Entrez Gene ID: (Human) 1982, (Mouse) 13690
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.