Novus Biologicals
Manufacturer Code:NBP216100
Catalog # NBP216100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human DAP3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: 28S ribosomal protein S29, mitochondrial; bMRP-10 DAP-3Death-associated protein 3 death associated protein 3 DKFZp686G1215928S ribosomal protein S29 mitochondrial FLJ12817 MGC126058 MGC126059 mitochondrial 28S ribosomal protein S29 MRP-S29Ionizing radiation resistance conferring protein MRPS29S29mt; DAP-3; Death-associated protein 3; Ionizing radiation resistance conferring protein; mitochondrial 28S ribosomal protein S29; Mitochondrial small ribosomal subunit protein mS29; MRP-S29; S29mt
Gene Aliases: bMRP-10; DAP-3; DAP3; MRP-S29; MRPS29
UniProt ID: (Human) P51398
Entrez Gene ID: (Human) 7818
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.