Proteintech
Manufacturer Code:102761AP150UL
Catalog # 50-172-6172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | DAP3 Fusion Protein Ag0274 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28S ribosomal protein S29, mitochondrial; bMRP 10 DAP 3 DAP3 death associated protein 3 MRP S29 MRPS29 S29mt; DAP-3; Death-associated protein 3; Ionizing radiation resistance conferring protein; mitochondrial 28S ribosomal protein S29; Mitochondrial small ribosomal subunit protein mS29; MRP-S29; S29mt
Gene Aliases: 4921514D13Rik; bMRP-10; DAP-3; DAP3; MRP-S29; MRPS29; S29mt
UniProt ID: (Human) P51398, (Mouse) Q9ER88
Entrez Gene ID: (Human) 7818, (Mouse) 65111
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.