R&D Systems
Manufacturer Code:IC5240T100UG
Catalog # IC5240T100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 406288 |
Immunogen | Human Dap12 synthetic peptide QGQRSDVYSDLNTQRPYYK Accession # O43914 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: DAP12PLOSL DNAX-activation protein 12 KARAP KARAPPLO-SL KAR-associated protein killer activating receptor associated protein Killer-activating receptor-associated protein TYRO protein tyrosine kinase binding protein TYRO protein tyrosine kinase-binding protein TYROBP; DNAX-activation protein 12; KAR-associated protein; Killer-activating receptor-associated protein; TYRO protein tyrosine kinase-binding protein
Gene Aliases: DAP12; KARAP; PLOSL; TYROBP
UniProt ID: (Human) O43914
Entrez Gene ID: (Human) 7305
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.