Invitrogen
Manufacturer Code:PA5109615
Catalog # PIPA5109615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-102 of human DAP (NP_004385.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4921531N22Rik AI789753 AU040360 Dap DAP 1 DAP1 DAP-1 Death-associated protein 1 MGC99796 OTTHUMP00000161670 OTTHUMP00000221331 Rap7a; DAP-1; Death-associated protein 1; Rap7a
Gene Aliases: 4921531N22Rik; AI789753; AU040360; DAP; DAP1; Rap7a
UniProt ID: (Human) P51397, (Mouse) Q91XC8, (Rat) Q9QX67
Entrez Gene ID: (Human) 1611, (Mouse) 223453, (Rat) 64322
Molecular Function: enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.